Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Ethylenediaminetetraacetic acid, tripotassium salt dihydrate, 99+%

Catalog No. AC301600025
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC301600025 2.5 kg Plastic bottle
AC301601000 100 g Plastic bottle
AC301605000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC301600025 Supplier Thermo Scientific Chemicals Supplier No. 301600025
Add to Cart

CAS: 65501-24-8 | C10H13K3N2O8·2H2O | 442.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65501-24-8
Molecular Formula C10H13K3N2O8·2H2O
Molecular Weight (g/mol) 442.56
InChI Key MAPFUJCWRWFQIY-UHFFFAOYSA-K
Synonym ethylenediaminetetraacetic acid tripotassium salt dihydrate, edta tripotassium salt, edta tripotassium salt dihydrate, tripotassium ethylenediaminetetraacetate dihydrate, potassium 2,2'-2-carboxylatomethyl carboxymethyl amino ethyl azanediyl diacetate dihydrate, tripotassium 2-2-bis carboxylatomethyl amino ethyl carboxymethyl amino acetate dihydrate, edta tripotassium salt, dihydrate, ethylenediaminetetraacetic acid, tripotassium salt dihydrate, ethylenediaminetetraacetic acid tripotassium salt dihydrate titration
PubChem CID 2733162
IUPAC Name tripotassium;2-[2-[bis(carboxylatomethyl)amino]ethyl-(carboxymethyl)amino]acetate;dihydrate
SMILES C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)O)CC(=O)[O-].O.O.[K+].[K+].[K+]

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 99% min. (on dry substance) (Complexometry)
Packaging Plastic bottle
Linear Formula (KOCOCH2)2NCH2CH2N(CH2COOK)CH2COOH·2H2O
Quantity 2.5 kg
Formula Weight 442.56
Percent Purity 99+%
Physical Form Fine Crystalline Powder
Chemical Name or Material Ethylenediaminetetraacetic acid, tripotassium salt dihydrate
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.