Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ethylmagnesium bromide, 3M in diethyl ether, AcroSeal™, Acros Organics
Click to view available options
Quantity:
100mL
800mL
Packaging:
AcroSeal™ Glass bottle
Specifications
Color | Brown |
Flash Point | −40°C |
Specific Gravity | 1.03 |
Quantity | 100mL |
Density | 1.0300g/mL |
Packaging | AcroSeal™ Glass bottle |
Linear Formula | C2H5MgBr |
Fieser | 02,205 |
Formula Weight | 133.27 |
Physical Form | Liquid |
Show More |
Safety and Handling
GHS H Statement
Extremely flammable liquid and vapour.
In contact with water releases flammable gases which may ignite spontaneously.
Harmful if swallowed.
Causes severe skin burns and eye damage.
May cause drowsiness or di
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove conta
GHS Signal Word: Danger
EINECSNumber : 213-127-3
TSCA : TSCA