Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylmagnesium bromide, 3M in diethyl ether, AcroSeal™

Catalog No. p-130944
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC347291000 100 mL AcroSeal™ Glass bottle
AC347298000 800 mL AcroSeal™ Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC347291000 Supplier Thermo Scientific Chemicals Supplier No. 347291000
Only null left
Add to Cart

CAS: 925-90-6 | C2H5BrMg | 133.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 925-90-6 , 60-29-7
Molecular Formula C2H5BrMg
Molecular Weight (g/mol) 133.27
MDL Number MFCD00000043
InChI Key TWTWFMUQSOFTRN-UHFFFAOYSA-M
Synonym ethylmagnesium bromide, ethyl magnesium bromide, magnesium, bromoethyl, ethylmagnesiumbromide, bromoethylmagnesium, ethylmagnesium bromide solution, 1.0 m in thf, ethylmagnesium bromide solution, 3.0 m in diethyl ether, etmgbr, ethylmagnesiumbromid, ethylmagnesium bromid, grignard reagent
PubChem CID 101914
IUPAC Name magnesium;ethane;bromide
SMILES CC[Mg]Br

Specifications

Color Brown
Density 1.0300g/mL
Flash Point −40°C
Packaging AcroSeal™ Glass bottle
Linear Formula C2H5MgBr
Quantity 100 mL
Fieser 02,205
Specific Gravity 1.03
Formula Weight 133.27
Physical Form Liquid
Chemical Name or Material Ethylmagnesium bromide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Extremely flammable liquid and vapour.
In contact with water releases flammable gases which may ignite spontaneously.
Harmful if swallowed.
Causes severe skin burns and eye damage.
May cause drowsiness or di
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove conta
EINECSNumber 213-127-3
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.