Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Ethylmercurithiosalicylic acid, sodium salt, 97.0-101.0%

Catalog No. AC118685000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC118685000 500 g Glass bottle
AC118680050 5 g Glass bottle
AC118680250 25 g Glass bottle
AC118681000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC118685000 Supplier Thermo Scientific Chemicals Supplier No. 118685000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Ethylmercurithiosalicylic acid, CAS # 54-64-8, is an oxidizing reagent that can inhibit various sulfhydryl-dependent enzymes and proteins. It is also known as thimerosal or 2-(ethylmercuriomercapto)benzoic acid sodium salt. ∣ CAS: 54-64-8 ∣ C9H9HgNaO2S ∣ 404.81 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Ethylmercurithiosalicylic acid is an oxidizing reagent that inhibits sulfhydryl-containing active sites of several enzymes
  • It is also known as thimerosal or 2-(ethylmercuriomercapto)benzoic acid sodium salt.
  • It can bind to sulfhydryl compounds, such as glutathione, cysteine, and sulfhydryl groups of proteins
  • Additionally, it can activate inositol triphosphate calcium channels on membranes of the endoplasmic reticulum, resulting in the regulation of cellular functions

Applications

  • Ethylmercurithiosalicylic acid has been widely investigated as a preservative in medicinal laboratory research, including vaccine studies
  • It has been shown to exhibit antifungal and antiseptic properties
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54-64-8
Molecular Formula C9H9HgNaO2S
Molecular Weight (g/mol) 404.81
MDL Number MFCD00013062
InChI Key RTKIYNMVFMVABJ-UHFFFAOYSA-L
Synonym thimerosal, thiomersal, thiomersalate, mercurothiolate, sodium merthiolate, sodium ethylmercurithiosalicylate, merthiolate, thimerosalate, thimerosalum, thimersalate
PubChem CID 16684434
ChEBI CHEBI:9546
IUPAC Name sodium;(2-carboxylatophenyl)sulfanyl-ethylmercury
SMILES [Na+].CC[Hg]SC1=CC=CC=C1C([O-])=O

Specifications

Melting Point 232.0°C to 233.0°C
Color White to Cream
pH 6 to 8 (1% soln.)
Physical Form Powder
Quantity 500 g
Flash Point 250°C
Infrared Spectrum Authentic
Loss on Drying 0.5% max. (vacuum/P2O5)
Packaging Glass bottle
Assay Percent Range 97.0-101.0%
Linear Formula 2-(C2H5HgS)C6H4CO2Na
Merck Index 15, 9470
Solubility Information Solubility in water: 1g/1mL (20°C). Other solubilities: 1g/8mL in alcohol, 1g/10mL in methanol, almost insoluble in ether and benzene
Formula Weight 404.8
Percent Purity 97.0 to 101%
Grade Ungraded
Chemical Name or Material Ethylmercurithiosalicylic acid, sodium salt
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
Fatal if swallowed.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove
EINECSNumber 200-210-4

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.