Learn More
Cayman Chemical ANTIBODY 2-MeCCPA 5mg
SDP
Supplier:  Cayman Chemical 309355 MG
Encompass_Preferred
An adenosine A1 receptor agonist; selective for adenosine A1 over A2A and A3 receptors in radioligand binding assays (Kis = 1.8, 3,900 and 5,000 nM, respectively); inhibits forskolin-induced activation of adenylyl cyclase in rat cortical membranes (IC50 = 13.1 nM); increases tail flick latency in the hot plate test and inhibits formalin-induced late nocifensive behavior in rats from 2.5-5 mg/kg
Catalog No. 50-297-8722
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.