Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam Etomidate, GABAA Agonist, Solid, >98% Purity, 10 mg
Description
- Agonist at GABAA receptors containing β³ subunits
- Molecular Weight: 244.29 Da
- Purity: >98%
- Anaesthetic and amnesic properties
- Form: Solid
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Substrate Properties | Chemical Substrate |
| Shipping Condition | Ambient |
| Target Molecule | GABAA Receptors (β3 subunits) |
| Form | Solid |
| Buffer | 100% Etomidate |
| Content And Storage | Store at ambient temperature. |
| Quantity | 10 mg |
| For Use With (Application) | Functional studies, GABAA receptor research |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?