Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam Etomidate, GABAA Agonist, Solid, >98% Purity, 50 mg
Description
- Molecular weight: 244.29 Da
- Purity: >98%
- Mechanism of action: Agonist at GABAA receptors containing β3 subunits
- Properties: Anaesthetic and amnesic
- Form: Solid
- Constituents: 100% Etomidate
- Storage: Store at Ambient
- Shipping: Ambient - Can Ship with Ice
Specifications
Specifications
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Target Molecule | GABAA Receptor (β3 Subunits) |
| Content And Storage | Store at ambient temperature. |
| Form | Solid |
| Buffer | 100% Etomidate |
| Quantity | 50 mg |
| For Use With (Application) | Functional studies |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?