Learn More
Selleck Chemical LLC Etomoxir sodium salt 25mg 828934-41-4
SDP
Supplier:  Selleck Chemical LLC S824425MG
Encompass_Preferred
Etomoxir sodium salt is an irreversible inhibitor of carnitine palmitoyltransferase-1 (CPT-1) on the outer face of the inner mitochondrial membrane. Etomoxir enhances palmitate-induced cell apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5843
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.