Learn More
Cayman Chemical (+)-Etomoxir (sodium salt), 828934-41-4, 25 mg
SDP
Supplier:  Cayman Chemical 1196925
Encompass_Preferred
Molecular Formula C15H18ClO4 • Na, Purity ≥98%, Formula Weight 320.7, Synonyms: (2R)-2-[6-(4-chlorophenoxy)hexyl]-2-oxiranecarboxylic acid monosodium salt, (R)-(+)-Etomoxir; An irreversible inhibitor of carnitine palmitoyltransferase 1, a mitochondrial enzyme involved in fatty acid -oxidation (IC50 = 5-20 nM in rat liver); also acts as a direct agonist of PPAR.
Catalog No. NC0961831
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.