Learn More
Enzo Life Sciences Etoposide (100mg). CAS: 33419-42-0
SDP
Supplier:  Enzo Life Sciences BMLGR3070100
Encompass_Preferred
Antitumor reagent. Topoisomerase II inhibitor. Induces apoptosis by FasL. Activates p53 resulting in upregulated expression of TRAIL-R2 (DR5) and Bak to overcome TRAIL resistance in Bax-deficient human colon carcinoma cancer cells. Alternative name: 4-Desmethylepipodophyllotoxin 9-(4,6-O-ethylidene)-beta-D-glucopyranoside, VP-16-213. Purity: ≥95% (HPLC). Solubility: Soluble in DMSO, dimethyl formamide, ethyl ether or chloroform:methanol (1:1); slightly soluble in methanol or acetone; insoluble in water. Long Term Storage: Ambient.
Catalog No. 50-201-0956
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.