Learn More
Apexbio Technology LLC GW501516 317318-70-0 100mg
SDP
Supplier:  Apexbio Technology LLC A4309100
Encompass_Preferred
GW501516 is a subtype-selective synthetic agonist targeting peroxisome proliferator-activated receptor delta (PPAR ) exhibiting a Ki value of approximately 1 1 nM PPAR widely expressed in various tissues mediates cholesterol metabolism and interacts with retinoid X receptor (RXR) In cellular reporter assays utilizing a GAL4-driven system GW501516 activates PPAR -dependent transcription with an EC50 value close to 1 2 nM and demonstrates roughly 1000-fold greater selectivity toward PPAR compared to other PPAR isoforms Experimental studies in primate models of metabolic disease have shown GW501516 promotes HDL-cholesterol elevation reduces fasting triglycerides modifies LDL/VLDL particle profiles to larger forms enhances cholesterol efflux via increasing ABCA1 expression and partially reduces hyperinsulinemia without adverse effects on glucose handling GW501516 serves as a research tool for exploring the mechanisms and functions of PPAR activation in metabolic regulation
Catalog No. 50-000-61421
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.