Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Etoposide Inhibitor
SDP

Catalog No. NB1226 Shop All Novus Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1226 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1226 Supplier Novus Biologicals™ Supplier No. NB1226

missing translation for 'validMainframeMsg'

Low molecular weight, organic compound that may help regulate biological process.

Biological Activity: Topoisomerase II inhibitor (IC50 = 59.2uM)
Storage: Store at 4°C.

Note: Inhibitors are guranteed for 1 year from date of receipt.

Specifications

Color White
Product Type Inhibitor
Target Proteins Etoposide
CAS 33419-42-0
Molecular Formula C29H32O1
Formula Weight 588.86g/mol
Solubility Soluble to 100mM in DMSO
Content And Storage Store at 4°C
For Use With (Application) Induces DNA breaks
Quantity 100 mg

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.