Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Etoposide, MP Biomedicals

Catalog No. ICN19391880 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19391880 100 mg
ICN19391825 25 mg
ICN19391883 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN19391880 Supplier MP Biomedicals, Inc Supplier No. 0219391880
Add to Cart
Add to Cart

Chemical Identifiers

CAS 33419-42-0
Molecular Formula C29H32O13
Molecular Weight (g/mol) 588.56
MDL Number MFCD00869325,MFCD00869325
InChI Key VJJPUSNTGOMMGY-MRVIYFEKSA-N
Synonym vjjpusntgommgy-nzlmilqcsa
PubChem CID 50936917
SMILES COC1=CC(=CC(OC)=C1O)[C@H]1[C@@H]2[C@H](COC2=O)[C@H](O[C@@H]2O[C@@H]3CO[C@@H](C)O[C@H]3[C@H](O)[C@H]2O)C2=CC3=C(OCO3)C=C12

Specifications

Color White
Quantity 100 mg
IUPAC Name (9R)-5-[[(2R,6R,7R,8R)-7,8-dihydroxy-2-methyl-4,4a,6,7,8,8a-hexahydropyrano[3,2-d][1,3]dioxin-6-yl]oxy]-9-(4-hydroxy-3,5-dimethoxyphenyl)-5a,6,8a,9-tetrahydro-5H-[2]benzofuro[6,5-f][1,3]benzodioxol-8-one
Formula Weight 588.6
Physical Form Powder
Chemical Name or Material Etoposide
EINECSNumber 251-509-1
Recommended Storage Store at Room Temperature (15 to 30°C).
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.