Learn More
Trajan™ ETP Active Film™ Electron Multipliers
SDP
Catalog No. SG14820 Shop All SGE Inc. Products
Change view
Click to view available options
For Use With (Equipment):
Comstock MiniTOF TOF Analyzer Type MS
Kratos (Shimadzu) Kompact MALDI (I-IV) and Various Bruker TOF Analyzer Type MS Instruments
Sensar/Larson-Davis TOF 2000 TOF Analyzer Type MS
Thermo Elemental PQ (SXP Rods), PQ-3, Excel (Sequential) Quadrupole Analyzer Type ICPMS
Varian UltraMass Quadrupole Analyzer Type ICPMS
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
SG14820 Kratos (Shimadzu) Kompact MALDI (I-IV) and Various Bruker TOF Analyzer Type MS Instruments
SG14824 Comstock MiniTOF TOF Analyzer Type MS
SG14562A Thermo Elemental PQ (SXP Rods), PQ-3, Excel (Sequential) Quadrupole Analyzer Type ICPMS
SG14823H Sensar/Larson-Davis TOF 2000 TOF Analyzer Type MS
SG14566 Varian UltraMass Quadrupole Analyzer Type ICPMS
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. SG14820 Supplier Trajan™ Supplier No. 14820
Drop-in, plug-compatible replacement electron multipliers
  • Detect presence of ion signals emerging from the mass analyzer of mass spectrometer; also reduce multiplier noise and increase signals
  • Available for all major instruments including Agilent Technologies™ Perkin-Elmer™, Shimadzu™, Thermo Scientific, and Varian™; total compatibility with GC/MS, LC/MS, ICP/MS, TOF/MS, and Magnetic Sector techniques
  • Features Active Film™—proprietary dynode material with a chemically durable metal oxide secondary emission system; very air stable; exhibits stable performance even after repeated exposure to atmosphere
  • Large dynode surface area for extended operating life
  • High operational reliability and mechanical robustness; designed for sensitivity and linear system response
  • Optimized ion and electron optics for maximum performance
  • Easy installation
  • Meet EPA, DFTPP ion abundance criteria
  • Two-year shelf life
Specifications
Type Electron Multiplier
For Use With (Application) TOF/MS - Detect Presence of Ion Signals Emerging From The Mass Analyzer of Mass Spectrometer, Also Reduce Multiplier Noise and Increase Signals
For Use With (Equipment) Kratos (Shimadzu) Kompact MALDI (I-IV) and Various Bruker TOF Analyzer Type MS Instruments
Max. Temperature (Metric) 350°C
Surface Type Large Dynode
Certifications/Compliance EPA, DFTPP
Max. Temperature (English) 662°F