Learn More
Trajan™ ETP Active Film™ Electron Multipliers
SDP
Catalog No. SG14824 Shop All SGE Inc. Products
Change view
Click to view available options
For Use With (Equipment):
Comstock MiniTOF TOF Analyzer Type MS
Kratos (Shimadzu) Kompact MALDI (I-IV) and Various Bruker TOF Analyzer Type MS Instruments
Sensar/Larson-Davis TOF 2000 TOF Analyzer Type MS
Thermo Elemental PQ (SXP Rods), PQ-3, Excel (Sequential) Quadrupole Analyzer Type ICPMS
Varian UltraMass Quadrupole Analyzer Type ICPMS
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
SG14824 Comstock MiniTOF TOF Analyzer Type MS
SG14820 Kratos (Shimadzu) Kompact MALDI (I-IV) and Various Bruker TOF Analyzer Type MS Instruments
SG14562A Thermo Elemental PQ (SXP Rods), PQ-3, Excel (Sequential) Quadrupole Analyzer Type ICPMS
SG14823H Sensar/Larson-Davis TOF 2000 TOF Analyzer Type MS
SG14566 Varian UltraMass Quadrupole Analyzer Type ICPMS
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. SG14824 Supplier Trajan™ Supplier No. 14824
Drop-in, plug-compatible replacement electron multipliers
  • Detect presence of ion signals emerging from the mass analyzer of mass spectrometer; also reduce multiplier noise and increase signals
  • Available for all major instruments including Agilent Technologies™ Perkin-Elmer™, Shimadzu™, Thermo Scientific, and Varian™; total compatibility with GC/MS, LC/MS, ICP/MS, TOF/MS, and Magnetic Sector techniques
  • Features Active Film™—proprietary dynode material with a chemically durable metal oxide secondary emission system; very air stable; exhibits stable performance even after repeated exposure to atmosphere
  • Large dynode surface area for extended operating life
  • High operational reliability and mechanical robustness; designed for sensitivity and linear system response
  • Optimized ion and electron optics for maximum performance
  • Easy installation
  • Meet EPA, DFTPP ion abundance criteria
  • Two-year shelf life
Specifications
Product Type Electron Multiplier
For Use With (Equipment) Comstock MiniTOF TOF Analyzer Type MS
For Use With (Application) TOF/MS - Detect Presence of Ion Signals Emerging From The Mass Analyzer of Mass Spectrometer, Also Reduce Multiplier Noise and Increase Signals
Max. Temperature (English) 662°F
Max. Temperature (Metric) 350°C
Surface Type Large Dynode
Certifications/Compliance EPA, DFTPP