Learn More
Invitrogen™ ETS2 Recombinant Rabbit Monoclonal Antibody (HL2259)
Shop All Thermo Scientific Products

Description
ETS2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF
Specifications
Specifications
| Antigen | ETS2 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | HL2259 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene | ETS2 |
| Gene Accession No. | P15036, P15037 |
| Gene Alias | AU022856; avian erythroblastosis virus E26 (v-ets) oncogene homolog 2; E26 avian leukemia oncogene 2, 3' domain; ETS proto-oncogene 2, transcription factor; Ets2; Ets-2; ETS2IT1; oncogene ETS-2; protein C-ets-2; v-ets avian erythroblastosis virus E2 oncogene homolog 2; v-ets avian erythroblastosis virus E26 oncogene homolog 2; v-ets erythroblastosis virus E26 oncogene homolog 2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.