Learn More
Cytiva Ettan™ IPGphor™ Strip Holder Cleaning Solution
Catalog No. 45002497 Shop All Cytiva Products
Change view
Click to view available options
Volume (Metric):
1 L
Catalog No. Volume (Metric)
45-002-497 1 L
1 options
Catalog No. 45-002-497 Supplier Cytiva Supplier No. 29011927
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart
Strip Holder Cleaning Solution is designed for mild cleaning of ceramic stripholders used for isoelectric focusing of protein samples
IPGphor Strip Holder Cleaning Solution has been specifically formulated for removing protein deposits. Strip Holder Cleaning Solution is supplied in 1L bottle.
Compatible with these stripholders and trays used with Immobiline™ DryStrips:
  • IPGphor Manifold Ceramic Tray
  • IPGphor strip holders (various sizes)
  • IPGphor Cup Loading Manifold
  • IPGphor Manifold Light
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Designed for mild cleaning of Ceramic Ettan™ IPGphor™ strip holders, manifold Ceramic tray, cup loading manifold and manifold light used for isoelectric focusing of Protein samples
Volume (Metric) 1 L