Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ETV4 Monoclonal Antibody (OTI5C11), TrueMAB™, OriGene
SDP

Catalog No. 501684037
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-4037 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-4037 Supplier OriGene Supplier No. TA811432
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ETV4: ets variant gene 4 (E1A enhancer binding protein, E1AF), also known as PEA3. Several members of the Ets gene family are known to encode sequencespecific DNA binding proteins. These include mouse PU.1, mouse and human Ets-1, Drosophila E74, chicken and human Ets-2 and rat GABP-a. Each of these proteins recognizes similar motifs in DNA that share a centrally located 5′-GGAA-3′ element. For instance, PEA3 binds the motif 5′-AGGAAG-3′ (the PEA-3 motif), but does not bind to the sequence 5′-AGGAAC-3′, recognized by PU.1, although PU.1 binds equally well to both sequences. It appears thatall of the Ets proteins recognize the same central core sequence but that each protein interacts with unique sequences that flank this core. PEA3 is expressed at readily detectable levels in cells of epithelial and fibroblastic origin but is not expressed in hematopoietic cells. This is in contrast to other members of the Ets gene family, such as Ets-1, Ets-2 and Fli-1, each of whichis expressed primarily in cells of hematopoietic origin.

Specifications

Antigen ETV4
Applications Western Blot
Classification Monoclonal
Clone OTI5C11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene ETV4
Gene Accession No. P43268
Gene Alias E1A-F, E1AF, PEA3, PEAS3
Gene Symbols ETV4
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 1-276 of human ETV4 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2118
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.