Learn More
Enzo Life Sciences 5,8,11,14-Eicosatetraynoic acid (20mg). CAS: 1191-85-1
SDP
Supplier:  Enzo Life Sciences BMLET0040020
Encompass_Preferred
Inhibits arachidonic acid uptake. Inhibits cyclooxygenase (ID50=8 µM) and all lipoxygenases (ID50=10, 0.3, and 0.2 µM for 5-, 12- and 15-LO respectively) in whole cells. Inhibits PLA2 and cytochrome P-450. Modulates Ca2+ entry into cells. Stimulates luteinizing hormone release from cultured pituitary cells. PPAR agonist. Alternative name: ETYA. Purity: ≥98% (TLC). Solubility: Soluble in 100% ethanol, methanol, DMSO, acetone or acetonitrile. Long Term Storage: +4°C.
Catalog No. 50-201-0786
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.