Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eugenyl Acetate, MP Biomedicals

Catalog No. ICN212771 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN212771 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN212771 Supplier MP Biomedicals, Inc Supplier No. 0521277180

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 93-28-7
Molecular Formula C12H14O3
Molecular Weight (g/mol) 206.24
MDL Number MFCD00026191
InChI Key SCCDQYPEOIRVGX-UHFFFAOYSA-N
Synonym eugenyl acetate, acetyleugenol, eugenol acetate, aceteugenol, 4-allyl-2-methoxyphenyl acetate, acetyl eugenol, 1,3,4-eugenol acetate, 4-allyl-2-methoxyphenol acetate, aceto eugenol, 1-acetoxy-2-methoxy-4-allylbenzene
PubChem CID 7136
ChEBI CHEBI:34522
IUPAC Name 2-methoxy-4-(prop-2-en-1-yl)phenyl acetate
SMILES COC1=CC(CC=C)=CC=C1OC(C)=O

Specifications

Quantity 100 g
Formula Weight 206.26
Chemical Name or Material Eugenyl Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.