Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam EUK 134, SOD/Catalase Mimetic, >98% Purity, Solid, 5 mg

Catalog No. 502705128
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-5128 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-5128 Supplier Abcam Supplier No. AB1455935MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synthetic SOD/catalase mimetic with neuroprotective and anti-ischemic effects.

  • Molecular Weight: 416.7 Da
  • Purity: >98%
  • Form: Solid
  • Function: SOD/catalase mimetic
  • Activity: Consumes H2O2 with an initial reaction rate of 234 μM/min
  • Biological Effects: Ameliorates nNOSμ translocation and skeletal muscle fiber atrophy during short-term mechanical unloading; shows neuroprotective and anti-ischemic effects in vivo
  • Storage: Store at -20°C

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Substrate Properties Chemical Substrate
Substrate H2O2
Target Molecule H2O2
Shipping Condition Ambient
Form Solid
Content And Storage Store at -20°C.
Product Type Antioxidant
Buffer 100% manganese(2+);2-methoxy-6-[2-[(3-methoxy-2-oxidophenyl)methylideneamino]ethyliminomethyl]phenolate;chloride
Formulation 100% manganese(2+);2-methoxy-6-[2-[(3-methoxy-2-oxidophenyl)methylideneamino]ethyliminomethyl]phenolate;chloride
Quantity 5 mg
For Use With (Application) Neuroprotection, Anti-ischemia, Skeletal muscle fiber atrophy research
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.