Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ Eukaryotic 18S rRNA Endogenous Control (FAM™/MGB probe, non-primer limited)
GREENER_CHOICE

Catalog No. 4352930E
Encompass_Preferred
Change view
Click to view available options
No. of Reactions:
125 Reactions
2500 Reactions
500 Reactions
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
43-529-30E 2500 Reactions
43-337-60T 125 Reactions
43-337-60F 500 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 43-529-30E Supplier Applied Biosystems™ Supplier No. 4352930E
  • $1,142.00
    $1,119.65 / Each of 1

    Save $22.35

    2% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Eukaryotic 18S rRNA Endogenous Control (FAM™/MGB probe, non-primer limited)

The Applied Biosystems™ Eukaryotic 18S rRNA Endogenous Control (FAM™ Dye⁄MGB Probe, Non-Primer Limited) is intended as an endogenous control. It allows relative gene expression quantification in cDNA samples when used with other gene expression assays. Probe is labeled with 6FAM™ dye - MGB and the primers are not limited. Can be used for singleplex PCR reactions. Endogenous control is to be used with Inventoried and Non-Inventoried TaqMan™ Gene Expression Assays, Custom TaqMan™ Gene Expression Assays, and⁄or Custom TaqMan™ Primers and Probes.

Assay Details:

Gene Symbol: 18S
RefSeq: X03205.1
Probe Exon Location: NA
Amplicon Size: 187
Corresponding TaqMan Assay ID: Hs99999901_s1

TaqMan™ Endogenous Controls

Eliminate months of assay design, formulation, and testing by using TaqMan™ Endogenous Controls as your controls to quantify gene expression. This convenient collection of pre-designed probe and primer sets enables you to normalize the amount of sample RNA or DNA in a reaction.

Complete Solution for Quantitative Gene Expression

Having a hard time deciding what controls to use to quantify gene expression — even with detailed information on biological systmes?Now, with TaqMan™ Endogenous Controls, you can avoid all the trial-and-error of selecting controls for most common human, mouse, rat, and eukaryotic genes.

Simple to Use

All components of the TaqMan™ Endogenous Controls are QC tested, formulated into a single 20X mix, and functionally tested. The controls are not only simple to use, but they are also fully compatible with universal conditions for two-step RT-PCR. Just add TaqMan™ Universal PCR Master Mix (with or without AmpErase™ UNG) and your cDNA sample to generate sensitive, reproducible, and truly quantitative gene expression data on Applied Biosystems instruments including the Applied Biosystems 7900HT, 7300, 7500 Real-Time PCR Systems, and the 7000 and 7700 Sequence Detection Systems.

Flexible Offering

We build each endogenous control using our proven 5' nuclease chemistry. For maximum flexibility, you can choose between two different reporter dyes and two quenchers:
  • A FAM™ dye-labeled TaqMan™ MGB probe (250nM, final concentration) and two unlabeled PCR primers (900nM each)
  • A VIC™ dye-labeled TaqMan™ MGB probe (250nM, final concentration) and two unlabeled PCR primers (150nM each y primer limited)
  • A VIC™ dye-labeled TAMRA™ probe (250nM, final concentration) and two unlabeled PCR primers (150nM each y primer limited)

    Choosing the Right Endogenous Control

    Endogenous controls can normalize the expression levels of target genes by correcting differences in the amount of cDNA that is loaded into PCR reaction wells. For best results, verify that the endogenous control is consistently expressed in the sample set to be tested. Endogenous control expression must be uniform across all samples in the study. For multiplexing, ensure that the gene expression level of the endogenous control is greater than that of the target.

    Multiplex vs. Singleplex PCR

    All TaqMan™ Endogenous Controls that contain probes labeled with the VIC™ reporter dye are primer limited. This allows multiplexing of TaqMan™ Endogenous Controls with target gene expression assays, provided that the control gene is more abundantly expressed than the target gene. All TaqMan™ Endogenous Controls that contain probes labeled with the FAM™ reporter dye are not primer limited and are not intended for multiplexing .

    For Research Use Only. Not for use in diagnostic procedures.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Concentration 20X
    Accession Number X03205.1
    Amplicon Size 187
    Content And Storage Control Primer-Probe Set(s)
    Detection Method Primer-probe
    Format Tube
    PCR Method 2-step RT-qPCR
    Gene Symbol 18S
    Label or Dye FAM
    Product Line FAM, TaqMan
    Quantity 2500 reactions
    Shipping Condition Room Temperature
    Species Eukaryotic, Rat, Mouse, Human
    Target 18S rRNA
    For Use With (Application) Gene Expression
    No. of Reactions 2500 Reactions
    Show More Show Less
    I am working on both mouse and human samples for gene expression studies. Do you have one endogenous control that I can use with both species?

    The 18S rRNA is highly conserved among eukaryotes. We have several configurations of assays for this gene: P/N 4331182, Assay ID: Hs99999901_s1; PN 4319413E, 4310893E, 4333760T, 4333760F, 4308329. You can use any of these for both human and mouse samples, as well as samples from any other eukaryotic species.

    What are the key features of a TaqMan MGB probe?

    TaqMan MGB probes have a nonfluorescent quencher at the 3´ end and a minor groove binder at the 3´ end that increases the melting temperature (Tm) of probe, allowing the use of shorter probes.  For more information on MGB probes, please consult the following manual: "Primer Express v1.5 and TaqMan MGB Probes for Allelic Discrimination: All PCR Instruments: User Bulletin." You can find a copy on our website by entering this title in the main search box.

    When using the TaqMan Gene Signature Plates, should I mix the reactions after adding the master mix and template to the wells containing lyophilized primers and probe?

    No, but it is recommended that the plate be centrifuged to draw down the liquid to the bottom of the wells.

    What are your recommendations for the best way to dilute and store my TaqMan probe?

    The probe should be diluted in TE buffer [10 mM Tris-HCl, pH 8.0 (at 25°C), 1 mM EDTA]. We recommend storing your probes, both stocks and working solutions, at -20°C in aliquots. This is to cut down on the number of times the probe is subject to freeze-thaws. For detailed information on diluting primers and probes, please refer to the tutorial entitled "Reconstituting and Diluting Primers and TaqMan Probes". You can find it on our website by entering this title in the main search field.

    WARNING: Cancer - www.P65Warnings.ca.gov

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.