Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Euramco Hazardous Location Anti-Static Ducts
SDP

Catalog No. 19817743
Change view
Click to view available options
Length (English):
15 ft.
25 ft.
Length (English) Duct:
180 in.
300 in.
Weight (English):
9 lb.
16 lb.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English) Length (English) Duct Weight (English)
19-817-743 25 ft. 300 in. 16 lb.
19-817-741 15 ft. 180 in. 9 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-817-743 Supplier Euramco Supplier No. FDT0825CBB

missing translation for 'validMainframeMsg'

Safely conducts electricity to the ground circuit of the blower

  • Non-collapsible and reinforced
  • Flame retardant
  • Surface resistance less than 6 GΦ

Specifications

Color Black
Diameter (English) Fan 8 in.
Diameter (Metric) Fan 20.32 cm
Product Type Anti-static Duct
Length (English) 25 ft.
Length (English) Duct 300 in.
Length (Metric) 7.6 m
Length (Metric) Duct 762 cm
Material Conductive PVC
Weight (English) 16 lb.
Weight (Metric) 7.25 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.