Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Euramco Ramfan™ EFi150XX Blower/Exhauster
SDP

Catalog No. 19822706
Change view
Click to view available options
Motor: 1.5Hp/1.1Kw; 115/230VAC, 50/60Hz
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-822-706 Motor: 1.5Hp/1.1Kw; 115/230VAC, 50/60Hz
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-822-706 Supplier Euramco Supplier No. EG8200XX

missing translation for 'validMainframeMsg'

For safe use in adverse atmospheres

  • High-strength
  • Lightweight anti-static glass reinforced ABS housing
  • IP55 rain tested switch enclosure
  • For hazardous or explosive gases safety

Certifications

ATEX, UL.

Specifications

Color Black
Depth (English) 16 in.
Depth (Metric) 40.64 cm
Certifications/Compliance ATEX, UL
Diameter (English) Fan 16 in.
Amperage 1.87 A
Diameter (Metric) Fan 40.64 cm
Product Type Blower/Exhauster
Voltage 115/230 V
Electrical Requirements 115/230 V, 50/60 Hz
Height (English) 19 in.
Horsepower 1.5 hp
Height (Metric) 48.26 cm
Material Conductive ABS
Noise 90.2 dB
Width (Metric) 45.72 cm
Weight (English) 61 lb.
Weight (Metric) 27.66 kg
Width (English) 18 in.
Frequency 50/60 Hz
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.