Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Europium(II) bromide, 99.99% (metals basis)

Catalog No. AA4443114
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4443114 25 g
AA4443103 1 g
AA4443106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4443114 Supplier Thermo Scientific Chemicals Supplier No. 04443114
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 13780-48-8 | Br2Eu | 311.772 g/mol

Europium(II) bromide is used in water treatment, chemical analysis and in certain crystal growth applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13780-48-8
Molecular Formula Br2Eu
Molecular Weight (g/mol) 311.772
MDL Number MFCD01076548
InChI Key PJVPGMOCWCUQHP-UHFFFAOYSA-L
Synonym europium dibromide, europium ii bromide, europium ii ; bromide, acmc-1bp16, europium ii bromide trace rare earth metals basis 1g
PubChem CID 53249294
IUPAC Name europium(2+);dibromide
SMILES [Br-].[Br-].[Eu+2]

Specifications

Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.99% (Metals basis)
Solubility Information Soluble in water.
Formula Weight 311.77
Odor Odorless
Sensitivity Hygroscopic
Chemical Name or Material Europium(II) bromide
Hazard Category H315-H319
Hazard Statement In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 13780-48-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.