Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Europium(III) chloride, ultra dry, 99.99% (REO)

Catalog No. p-7050157
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3573803 1 g
AA3573806 5 g
AA3573814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3573803 Supplier Thermo Scientific Chemicals Supplier No. 03573803
Only null left
Add to Cart

CAS: 10025-76-0 | Cl3Eu | 258.31 g/mol

Europium(III) chloride is used as a starting material for the preparation of other europium salts. It is also used in preparation of europium(II)chloride by reduction in a gold boat using hydrogen gas. Further, it is also used to prepare organometallic compounds of europium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-76-0
Molecular Formula Cl3Eu
Molecular Weight (g/mol) 258.31
MDL Number MFCD00010994
InChI Key NNMXSTWQJRPBJZ-UHFFFAOYSA-K
Synonym acmc-20akvq, europium 3+ trichloride, europium 3+ ion trichloride, europium iii chloride, ultra dry reo, europium chloride, anhydrous, europium iii chloride, anhydrous, ampuled under argon trace rare earth metals basis 1g
PubChem CID 9859933
SMILES [Cl-].[Cl-].[Cl-].[Eu+3]

Specifications

Melting Point 850°C
Physical Form Powder
Quantity 1 g
Merck Index 14,3904
Solubility Information Soluble in water; Soluble in dilute hydrochloric acid,acetone and alcohol; Slightly soluble in tetrahydrofuran.
IUPAC Name europium(3+);trichloride
Formula Weight 258.32
Percent Purity 99.99%
Packaging Ampule
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Europium(III) chloride, Ultra dry
Show More Show Less
EINECSNumber 233-040-4
RTECSNumber LE7525000
TSCA Yes
storageNote1
  • Ampouled under argon
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.