Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ EV Precipitation Solution (Urine)
SDP

Catalog No. NB397874
Change view
Click to view available options
Quantity:
30 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB397874 30 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB397874 Supplier Novus Biologicals™ Supplier No. NBP311766

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Specifications

Product Type EV isolation, exosome isolation, extracellular vesicle isolation
Quantity 30 mL
Sufficient For A 30mL solution is equalivant to 25 reactions (urine)
Content And Storage Store at room temperature.
Sample Type Urine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.