Description
- Uniquely designed, streamlined form makes PAPR lightweight, eliminates bulky corners that can dig into the wearer's back, and balances the unit's weight for maximum comfort
- Easy-to-read fuel gauge displays remaining battery capacity at a glance
- Battery status can be checked at any time during use
- Bullard Active Flow Technology automatically responds to a worker's need for more or less air flow
- Two distinct audible alarms notify wearers of low battery and low air flow
- Alarms are designed so the sound of the alarm travels through the breathing tube directly to the wearer's ear
- Powered by a long-lasting 10,000-hour brushless motor
- Single filter design for easy use
- Filter locking tab notifies the user that the filter is in place with both a visual indication and audible click
Order Info
A NIOSH approved EVA PAPR system consists of the following properly-assembled components: (1) hood or mask; (2) breathing tube; (3) blower assembly; (4) purifying filter cartridge (sold separately). Check model system components carefully before ordering.
Compliance
NIOSH approved.
Specifications
Specifications
| Battery Life | 7 to 8 hours |
| Belt Material | Foam |
| Facepiece/Headtop Model | Double bib hood, ratchet suspension, quick-attach, taped seams |
| Includes | PAPR system, EVA blower, battery, charger, breathing tube |
| Material (Lens) | Acetate |
| Model | EVA20TICST |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.