Learn More
Apexbio Technology LLC Evacetrapib (LY2484595), 10mg. Cas: 1186486-62-3 MFCD: MFCD23105886. CETP inhibitor,potent and selective
SDP
Supplier:  Apexbio Technology LLC A437710
Encompass_Preferred
Evacetrapib is a potent and selective inhibitor of cholesteryl ester transfer protein (CETP) with IC50 value of 5.5nM [1].As a benzazepine-based inhibitor of CETP, evacetrapib is developed to increase HDL cholesterol for the treatment of coronary artery disease. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4934