Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Evans Blue

Catalog No. AC195550250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC195550250 25 g Glass bottle
AC195550050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC195550250 Supplier Thermo Scientific Chemicals Supplier No. 195550250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

CAS: 314-13-6 | C34H24N6Na4O14S4 | 960.79 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 314-13-6
Molecular Formula C34H24N6Na4O14S4
Molecular Weight (g/mol) 960.79
MDL Number MFCD00004021
InChI Key KBNIFDASRCWYGC-GXNXWABVSA-J
Synonym C.I. 23860, Direct Blue 53
PubChem CID 24832074
IUPAC Name tetrasodium (6E)-4-amino-6-[2-(4'-{2-[(2E)-8-amino-1-oxo-5,7-disulfonato-1,2-dihydronaphthalen-2-ylidene]hydrazin-1-yl}-3,3'-dimethyl-[1,1'-biphenyl]-4-yl)hydrazin-1-ylidene]-5-oxo-5,6-dihydronaphthalene-1,3-disulfonate
SMILES [Na+].[Na+].[Na+].[Na+].CC1=CC(=CC=C1N\N=C1/C=CC2=C(C=C(C(N)=C2C1=O)S([O-])(=O)=O)S([O-])(=O)=O)C1=CC=C(N\N=C2/C=CC3=C(C=C(C(N)=C3C2=O)S([O-])(=O)=O)S([O-])(=O)=O)C(C)=C1

Specifications

Packaging Glass bottle
Solubility Information Solubility in water: 280g/L (20°C). Other solubilities: soluble in alcohol, acids, alkalies, insoluble in ether
Quantity 25 g
Formula Weight 960.78
Chemical Name or Material Evans Blue

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.