accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaNaNaH3CNHNNH2OSOOOSOOOHNNNH2OSOOOSOOOCH3(E)(E)

CAS RN 314-13-6

IUPAC Name: tetrasodium (6E)-4-amino-6-[2-(4'-{2-[(2E)-8-amino-1-oxo-5,7-disulfonato-1,2-dihydronaphthalen-2-ylidene]hydrazin-1-yl}-3,3'-dimethyl-[1,1'-biphenyl]-4-yl)hydrazin-1-ylidene]-5-oxo-5,6-dihydronaphthalene-1,3-disulfonate
Synonyms: evans blue (dye)
Molecular Weight (g/mol): 960.79
Molecular Formula: C34H24N6Na4O14S4
InChi Key: KBNIFDASRCWYGC-GXNXWABVSA-J
SMILES: [Na+].[Na+].[Na+].[Na+].CC1=CC(=CC=C1N\N=C1/C=CC2=C(C=C(C(N)=C2C1=O)S([O-])(=O)=O)S([O-])(=O)=O)C1=CC=C(N\N=C2/C=CC3=C(C=C(C(N)=C3C2=O)S([O-])(=O)=O)S([O-])(=O)=O)C(C)=C1
Molecular Weight (g/mol) 
  • (10)
  • (3)
Quantity 
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (1)
  • (4)
Color 
  • (4)
  • (1)
Grade 
  • (1)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (3)
  • (4)
  • (1)
  • (4)
  • (1)
  • (10)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (1)
  • (4)
  • (4)
  • (1)
  • (1)
  • (4)
  • (1)