Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Evans blue, MP Biomedicals™

Catalog No. icn15110805 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
100 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15110805 5 g
ICN15110810 10 g
ICN15110850 50 g
ICN15110880 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN15110805 Supplier MP Biomedicals, Inc Supplier No. 0215110805
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 314-13-6
Molecular Formula C34H24N6Na4O14S4
Molecular Weight (g/mol) 960.79
MDL Number MFCD00004021
InChI Key KBNIFDASRCWYGC-GXNXWABVSA-J
Synonym Direct blue 53
PubChem CID 24832074
IUPAC Name tetrasodium (6E)-4-amino-6-[2-(4'-{2-[(2E)-8-amino-1-oxo-5,7-disulfonato-1,2-dihydronaphthalen-2-ylidene]hydrazin-1-yl}-3,3'-dimethyl-[1,1'-biphenyl]-4-yl)hydrazin-1-ylidene]-5-oxo-5,6-dihydronaphthalene-1,3-disulfonate
SMILES [Na+].[Na+].[Na+].[Na+].CC1=CC(=CC=C1N\N=C1/C=CC2=C(C=C(C(N)=C2C1=O)S([O-])(=O)=O)S([O-])(=O)=O)C1=CC=C(N\N=C2/C=CC3=C(C=C(C(N)=C3C2=O)S([O-])(=O)=O)S([O-])(=O)=O)C(C)=C1

Specifications

Color Black
Quantity 5 g
Physical Form Powder
Formula Weight 960.82
Percent Purity 95%
Grade Not Available
Chemical Name or Material Evans blue, Dye content: ∽85%
EINECSNumber 206-242-5
Recommended Storage Store at Room Temperature (15 to 30°C). Store Desiccated.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.