Specifications
| Lens Type | H2X |
| Certifications/Compliance | ANSI Z87.1 |
| Lens Tint | Dark Gray |
| Frame Color | Black |
| Weight (Metric) | 22 g |
| Lens Material | Polycarbonate |
| Frame Material | Polycarbonate |