Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caplugs™ Evergreen Scientific False-Bottom Serum Tubes

Catalog No. 22046477 Shop All Caplugs Products
Change view
Click to view available options
Material:
Polystyrene
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
22-046-477 Polystyrene
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-046-477 Supplier Caplugs™ Supplier No. 1271788010
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

For use with most high-volume analyzers

  • Measure 4 in.
  • Three versions available: transparent polystyrene, non-breakable polypropylene (including a new threaded tube version)
  • Nominal capacity of 5mL
  • Accept bar codes and fit into most racks for high-volume analyzers
  • Threaded serum tube is available with an HDPE screw cap (sold separately); may also be used as a transport and storage vial
  • After samples are drawn, they are accepted into LIS as received, then placed into the analyzer
  • For storage, place a screw cap on it and place it in the refrigerator; no need to transfer screw cap vial and relabel
  • Piggyback serum cups available in polystyrene

Specifications

Color Natural
Product Type Serum Tube
Material Polystyrene
For Use With (Equipment) Olympus AU-5000 and BMC/Hitachi™ 736/737 analyzers
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.