Learn More
Description
- Flange is inserted into evacuated blood tube and pushed downward
- Wide body style prevents sample probes from hitting side walls
- Also for use in removing fibrin and macro particles from serum to prevent line clogging
- Can be used with Evergreen serum tubes
Specifications
Specifications
| Capacity (Metric) | 9 mL |
| Length (Metric) | 107mm |
| Product Type | Serum Filter |
| Capacity | 0.30 oz., 9mL |
| For Use With (Application) | Removing fibrin and macro |
| Length (English) | 4.25 in. |
| For Use With (Equipment) | EverGreen Serum tubes |
| Quantity | 1000 Pack |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.