Learn More
Cayman Chemical IbrutInIb-d5 1mg
SDP
Supplier:  Cayman Chemical 225611 MG
Encompass_Preferred
A selective Mcl-1 inhibitor (Ki 490 nM) that disrupts the heterodimerization of Mcl-1/Bax and Mcl-1/Bak induces apoptosis of various pancreatic cancer cell lines (IC50s 3 4-16 1 M) demonstrates antitumor activity in a BxPC-3 xenograft mouse model
Catalog No. 50-298-6731
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.