Learn More
Abcam Everolimus, mTOR inhibitor, 25MG
Supplier:  Abcam AB14215125MG
Encompass_Preferred
MW 958.2 Da, Purity >98%. Potent mTOR inhibitor (IC₅₀ = 1.6 - 2.4 nM). Immunosupressant and anticancer activity in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6310
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.