Learn More
DueNorth Everyday G3 Winter Ice/Snow Traction Aid
Designed for running, walking or working while keeping you safe and secure in ice or snow. The Everyday G3 provides the grip that won’t let you slip.
Supplier:  DueNorth V3551270S/M
 View more versions of this product
Catalog No. 19-060-811
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
  • Slip Protection
  • 100% natural rubber stays strong and retains elasticity in subzero temperatures for maximized durability
  • Easy to take on and off
  • Easily adjustable
  • Replaceable military grade tungsten carbide spikes
  • Twelve pair of Ice Diamond spikes per pair
  • Molded designs ensure comfortable ergonomically superior fit
  • Designed for running, walking or working while keeping your safe and secure
Specifications
Specifications
Pull-on
Outdoor Surfaces, Ice Surfaces, Running, Walking
Small/Medium
Reusable
Yes
Black
Vulcanized Rubber
General Purpose
Unisex
Yes
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.