Learn More
Fluotics Hamilton Star and Vantage Compatible P50, sterile, Tips in Quadrant only
Supplier:  Fluotics LHA10.F
Encompass
Tecan Evo/Fluent Compatible P10 filter/sterile Working Volume 10uL Tip Color Clear Hanging Rack LiHa/FCA Compare To 3004804 24 racks/case with 96 tips per rack
Catalog No. 50-004-35273