Learn More
Fluotics Tecan Evo/Fluent Compatible P10, filter/sterile
Supplier:  Fluotics LHA10CB.ST
Encompass
Tecan Evo/Fluent Compatible P10 sterile Working Volume 10uL Tip Color Black Hanging Rack LiHa/FCA Compare To 30104803 24 racks/case with 96 tips per rack
Catalog No. 50-004-35139