Learn More
Fluotics Tecan Evo/Fluent Compatible P10, filter/sterile
Supplier:  Fluotics LHAB10CBR.ST
Encompass
Tecan Evo/Fluent Compatible P10 sterile Working Volume 10uL Tip Color Black Single Stack base included LiHa/FCA Compare To 30104975 40 racks/case with 96 tips per rack
Catalog No. 50-004-35199