Learn More
Fluotics Tecan Evo/Fluent Compatible P1000, sterile
Supplier: Fluotics LHAB1000R.F
Tecan Evo/Fluent Compatible P1000 filter/sterile Working Volume 950uL Tip Color Clear Single Stack base included LiHa/FCA 40 racks/case with 96 tips per rack