Learn More
Apexbio Technology LLC Evodiamine, 100mg Cas# 518-17-2
SDP
Supplier:  Apexbio Technology LLC N1600100
Encompass_Preferred
Evodiamine is an indole alkaloid found in Evodia rutaescarpa. It acts as an anti-inflammatory compound; the mechanism of action may be inhibition of prostaglandin E2 synthesis or by cyclooxygenase 2 induction and NF-B activation.1 Evodiamine has been stu Other sizes are available. Please inqury us for quote.
Catalog No. 50-199-8568
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.