Learn More
Enzo Life Sciences Evodiamine (50mg). CAS: 518-17-2
SDP
Supplier:  Enzo Life Sciences ALX350330M050
Encompass_Preferred
A cell permeable, non-pungent TRPV1 antagonist (Ki=5.95µM in rat) that induces apoptosis in U937 cells. Inhibits nitric oxide (NO) production. Also inhibits tumor cell migration in vitro. Is effective against multidrug-resistant cancer cells. Inhibits angiogenesis. Alternative name: 8,13,13b,14-Tetrahydro-14-methylindolo[2'3'-3,4]pyrido[2,1-b]quinazolin-5-[7H]-one. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (5mg/ml; warm to 40°C). Long Term Storage: -20°C.
Catalog No. 50-200-8844
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.