Learn More
World Precision Instrument EVOM Auto V3 For TEER Measurement in 24M HTS Plate
SDP
Supplier:  World Precision Instrument EVAMT0303
Encompass
World Precision Instruments (WPI), the global leader in TEER technology, is proud to introduce the EVOM™ Auto, our newest entry into our pioneering EVOM family of products. The EVOM™ Auto automates measurements of TEER in epithelial or endothelial monolayers cultured on high throughput screening (HTS) 24 and 96-well and plates utilizing our innovative EVOM technology, qualitatively measuring cell monolayer health and quantitatively measuring cell confluence by determining an increase or a plateau in tissue resistance.  Automated measurement of tissue resistance of  transwell cell cultures provides the advantages of speed and precision, minimizes the chances of contamination, and ensures the rapid availability of measured resistance data. EVOM™ Auto produces a low AC current that omits any adverse physiological effects of the measurement technique on the samples.
Catalog No. NC2819322
Only null left
Add to Cart
Request a Quote