Learn More
World Precision Instrument EVOM™ AUTOLCI, Brightfield, 10X Magnification, with Red Fluorescence
SDP
Supplier:  World Precision Instrument EVILCI0106
Encompass_Preferred
EVOM™ AutoLCI is an automated live cell imaging system that is equipped with an advanced fluorescence and bright field microscopy, auto-focusing and real-time, multi-position imaging technology for a well plate, dish or T-flask. The streamlined process provides an easy workflow solution giving you a full set of tools you need to acquire the best quality images and accurate research results. The compact nature of the EVOM™ AutoLCI allows positioning in an incubator, providing improved cell viability, because there are fewer disturbances over the course of your experiment, reducing chances of cellular abnormality.
Catalog No. 50-252-1186
Only null left
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.