Learn More
Strem, An Ascensus Company Evonik Noblyst P8078 10g
SDP
Supplier:  Strem, An Ascensus Company 78153610G
Encompass
CAS# 7440-06-4. 10g. Platinum 1% and vanadium 2%, on activated carbon (50-70% wetted powder) Evonik Noblyst P8078. MFCD16987555. Molecular Weight: . Molecular Formula: . Color/form: wetted, black pwdr. Strem# 78-1536. http://www.strem.com/catalog/v/78-1536/
Catalog No. 50-203-4214