Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EVOS™ EVOS™ 60X Objective, fluorite, LWD

Catalog No. 12563468
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-563-468 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-563-468 Supplier EVOS™ Supplier No. AMEP4626
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Components manufactured to ensure publication-quality images

RMS-threaded objectives with 45mm parfocal distance

TRUSTED_SUSTAINABILITY

Specifications

Type Objective Lens
Product Line EVOS
For Use With (Equipment) EVOS™ FL Color Imaging System, EVOS™ XL Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System
Includes Vessels, Slides, Cell Culture Dishes, Flasks, Microtiter Well Plates
Lens Type Plan Fluorite
Magnification 60X
Numerical Aperture 0.75
Quantity 1 Ea.
I'm using an EVOS imaging system and my objective is rubbing up against the edge of the vessel holder of my stage. How can I correct this?

Objectives can hit the vessel holder when they are focused too high in the Z axis (up and down). This is a particularly a problem with the EVOS FL Auto Imaging System during instrument start-up, when the stage moves during system initiation, or when changing objectives. Coverslip-corrected objectives tend to be wider and flatter at the top of the barrel, which means that they are more likely to run into the edges of the vessel holder, particularly if you are imaging at the edges of the sample container. In those cases, use of that objective for those areas of the container may not be possible. If the objective if “jammed” by the vessel holder, then carefully unscrew the thumbscrews of the vessel holder and lift it straight off the stage, then move the objective downward in focus and toward the center of the stage. It is a good idea to have a shut-down procedure in your lab that includes moving the objectives to the lowest magnification and focusing downward with course focus prior to turning off the instrument for the day.
An objective can be damaged by scraping against the vessel holder. If this happens, take out the objective and examine it carefully for damage, particularly on the lens.

My objective keeps running into my sample when I'm trying to focus with my EVOS FL Auto Imaging System. What can I do about this?

If the lens is running up into a sample, this may be an issue with either focusing too quickly and missing the focal plane (if focusing manually) or a problem with the objective calibration (if using autofocus). It is a good idea to calibrate your objectives using the FL Auto calibration slide that comes with the system. Check to see if your objective is a long-working distance (LWD) or coverslip-corrected objective (CC). If coverslip-corrected, it is only for use with very short working distances for imaging through thin coverslips, but not through the slide or through plastics in microplates or culture dishes). If working with high magnification and oil immersion, by eye, move the objective upwards to touch the bottom of the sample and then only move slowly away from the sample for further focusing. An objective lens can be seriously damaged by scraping against samples. If this happens, check the objective lens for damage.

The objectives available for the EVOS cell imaging systems come in different formats: Plan Achromat, Plan Fluorite, and Plan Apochromat. What factors should I consider when selecting an objective?

Plan Achromat objectives are perfect for general applications where color and focus have standard correction; these are suitable for samples requiring low magnification (2x to 4x). Plan Achromat objectives are recommended for basic brightfield microscopy and simple fluorescence detection.
Plan Fluorite objectives provide the next level of improved resolution for brighter fluorescence signal and high contrast. These objectives are recommended for basic fluorescence imaging and brightfield microscopy at higher magnifications.
Plan Apochromat provides the highest level of resolution, fluorescence brightness, contrast, and chromatic correction. If you are imaging very small structures and require high contrast and brightness, the Plan Apochromat objectives are the best option.

What is the difference between "long working distance" (LWD) objectives and "coverslip corrected" (CC) objectives in the EVOS imaging systems?

All the EVOS imaging systems are inverted microscopes. For CC objectives, the coverslip must be face down, facing the objectives as the lenses have a short working distance suitable only for thin glass or plastic coverslips. LWD objectives are designed for viewing from the bottom of microplates, petri dishes, or culture flasks; the longer working distances of the lenses in these objectives accommodate thicker materials such as the plastic bottoms of various vessels.

WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.