Learn More
Invitrogen™ EVOS™ Light Cube, CFP 2.0
Catalog No. 12563697
Change view
Click to view available options
Quantity:
1 each
Catalog No. Quantity
12-563-697 1 each
1 options
Catalog No. 12-563-697 Supplier Invitrogen™ Supplier No. AMEP4953
Add to Cart
Add to Cart
Request a Quote
EVOS LED light cubes provide superior illumination uniformity across the field of view, resulting in exceptional tile-stitch image quality. At the same time, optimized spectral fidelity across channels significantly improves data quality by minimizing undesired bleed-through.
  • Second-generation EVOS LED light cubes provide superior illumination uniformity across the field of view, resulting in exceptional tile-stitch image quality
  • At the same time, optimized spectral fidelity across channels significantly improves data quality by minimizing undesired bleed-through
  • Like the original design, they combine condensing optics and hard-coated filters into one light engine that delivers digitally controlled fluorophore excitation
  • These LED light cubes are backwards compatible with older EVOS and Countess systems. and can be used with first-generation light cubes
  • Like the original light cubes, these LED cubes are user-changeable and automatically recognized by the instrument
  • The EVOS Light Cube, CFP 2.0, is ideal for use with CellLight CFP, ECFP, Evans Blue, Lucifer Yellow, and other fluorophores with excitation and emission maxima near 445 and 510 nm
  • The LED bulbs on the EVOS systems are rated for >50,000 hours, compared to 300 hours for a typical mercury bulb and 1,500 hours for a metal halide bulb
EVOS LED light cubes offer these important advantages:
  • Uniform illumination across the entire field of view for superb image quality
  • Optimized spectral compatibility to maximize true signal detection, while minimizing bleed-through for the best possible data quality
  • A short light path that affords superior detection of fluorescent signals
  • Stable illumination intensity over 50,000 hours that helps experimental reproducibility
TRUSTED_SUSTAINABILITY
Specifications
Type LED Light Cube
Product Line EVOS
For Use With (Equipment) Countess™ III FL Automated Cell Counter, EVOS™ M5000 Imaging System, EVOS™ M7000 Imaging System
Includes Condensing Optics, Hard-coated Filters
Wavelength Ex: 445/45 Em: 510/42
Quantity 1 each
For Research Use Only. Not for use in diagnostic procedures.