Learn More
Invitrogen™ EVOS™ Condenser Slider, Pinhole
Catalog No. 12563596
Change view
Click to view available options
Quantity:
1 each
Catalog No. Quantity
12-563-596 1 each
1 options
Catalog No. 12-563-596 Supplier Invitrogen™ Supplier No. AMEPAS15
Add to Cart
Add to Cart
Adds contrast when imaging
  • Used to improve illumination under certain conditions
  • Due to variances in sample size, color and thickness, actual slider use may differ from suggestions
  • Intended for brightfield applications at all magnifications to enhance contrast
Pinhole condenser slider is included with the EVOS™ FL and FL Color imaging systems.
TRUSTED_SUSTAINABILITY
Specifications
Type Condenser Slider
Product Line EVOS
For Use With (Equipment) EVOS™ FL Color Imaging System, EVOS™ XL Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System
Includes Diffuser Slider, Pinhole Slider, Meniscus Sliders
Quantity 1 each
WARNING: Cancer - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.